MilliporeSigma
  • Loss of Mpdz impairs ependymal cell integrity leading to perinatal-onset hydrocephalus in mice.
Loss of Mpdz impairs ependymal cell integrity leading to perinatal-onset hydrocephalus in mice.
EMBO molecular medicine (2017-05-14)
Anja Feldner, M Gordian Adam, Fabian Tetzlaff, Iris Moll, Dorde Komljenovic, Felix Sahm, Tobias Bäuerle, Hiroshi Ishikawa, Horst Schroten, Thomas Korff, Ilse Hofmann, Hartwig Wolburg, Andreas von Deimling, Andreas Fischer
ABSTRACT
Hydrocephalus is a common congenital anomaly.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Pyrogallol Red, Suitable for use as a complexometric indicator