MilliporeSigma
  • Targeting Cellular Calcium Homeostasis to Prevent Cytokine-Mediated Beta Cell Death.
Targeting Cellular Calcium Homeostasis to Prevent Cytokine-Mediated Beta Cell Death.
Scientific reports (2017-07-19)
Amy L Clark, Kohsuke Kanekura, Zeno Lavagnino, Larry D Spears, Damien Abreu, Jana Mahadevan, Takuya Yagi, Clay F Semenkovich, David W Piston, Fumihiko Urano
ABSTRACT
Pro-inflammatory cytokines are important mediators of islet inflammation, leading to beta cell death in type 1 diabetes. Although alterations in both endoplasmic reticulum (ER) and cytosolic free calcium levels are known to play a role in cytokine-mediated beta cell death, there are currently no treatments targeting cellular calcium homeostasis to combat type 1 diabetes. Here we show that modulation of cellular calcium homeostasis can mitigate cytokine- and ER stress-mediated beta cell death. The calcium modulating compounds, dantrolene and sitagliptin, both prevent cytokine and ER stress-induced activation of the pro-apoptotic calcium-dependent enzyme, calpain, and partly suppress beta cell death in INS1E cells and human primary islets. These agents are also able to restore cytokine-mediated suppression of functional ER calcium release. In addition, sitagliptin preserves function of the ER calcium pump, sarco-endoplasmic reticulum Ca
Sigma-Aldrich
MDL 28170, ≥90% (TLC)
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film
Sigma-Aldrich
Pioglitazone