MilliporeSigma
The
Frontiers in plant science (2017-08-22)
Imène Hichri, Yordan Muhovski, Eva Žižková, Petre I Dobrev, Emna Gharbi, Jose M Franco-Zorrilla, Irene Lopez-Vidriero, Roberto Solano, André Clippe, Abdelmounaim Errachid, Vaclav Motyka, Stanley Lutts
ABSTRACT
Salinity threatens productivity of economically important crops such as tomato (
Sigma-Aldrich
Zeatin, suitable for plant cell culture, BioReagent, powder
Sigma-Aldrich
Maltose solution, BioReagent, for molecular biology, ~20% in H2O