MilliporeSigma
  • Inhibitory effect of hesperetin on α-glucosidase: Molecular dynamics simulation integrating inhibition kinetics.
Inhibitory effect of hesperetin on α-glucosidase: Molecular dynamics simulation integrating inhibition kinetics.
International journal of biological macromolecules (2017-03-23)
Yan Gong, Xiu-Yuan Qin, Yuan-Yuan Zhai, Hao Hao, Jinhyuk Lee, Yong-Doo Park
ABSTRACT
The α-glucosidase inhibitor is of interest to researchers due to its association with type-2 diabetes treatment. Hesperetin is a flavonoid with natural antioxidant properties. This paper presents an evaluation on the effects of hesperetin on α-glucosidase via inhibitory kinetics using a Molecular Dynamics (MD) simulation integration method. Due to the antioxidant properties of hesperetin, it reversibly inhibits α-glucosidase in a slope-parabolic mixed-type manner (IC
Sigma-Aldrich
1-Naphthalenesulfonic acid, >50%