MilliporeSigma
  • Tuning the sound speed in macroporous polymers with a hard or soft matrix.
Tuning the sound speed in macroporous polymers with a hard or soft matrix.
Soft matter (2017-06-08)
Artem Kovalenko, Martin Fauquignon, Thomas Brunet, Olivier Mondain-Monval
ABSTRACT
In this paper, we investigate the factors affecting the sound speed in air-filled macroporous polymer materials at ultrasound frequencies. Due to the presence of large proportion of gas, these porous materials present high compressibility and, as a consequence, low sound speed which may fall down to values as low as 40 m s
Sigma-Aldrich
1-Ethynyl-1-cyclohexanol, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
5-(N,N-Dimethyl)amiloride hydrochloride
Pricing and availability is not currently available.