MilliporeSigma
  • Improving ruthenium-based ECL through nonionic surfactants and tertiary amines.
Improving ruthenium-based ECL through nonionic surfactants and tertiary amines.
The Analyst (2017-06-15)
Stefanie Kirschbaum-Harriman, Axel Duerkop, Antje J Baeumner
ABSTRACT
The influence of surfactants and coreactants on Ru(bpy)
Sigma-Aldrich
2-(Dibutylamino)ethanol, 99%
Sigma-Aldrich
Triton X-100, laboratory grade