MilliporeSigma
Intact Transition Epitope Mapping (ITEM).
Journal of the American Society for Mass Spectrometry (2017-06-16)
Yelena Yefremova, Kwabena F M Opuni, Bright D Danquah, Hans-Juergen Thiesen, Michael O Glocker
ABSTRACT
Intact transition epitope mapping (ITEM) enables rapid and accurate determination of protein antigen-derived epitopes by either epitope extraction or epitope excision. Upon formation of the antigen peptide-containing immune complex in solution, the entire mixture is electrosprayed to translate all constituents as protonated ions into the gas phase. There, ions from antibody-peptide complexes are separated from unbound peptide ions according to their masses, charges, and shapes either by ion mobility drift or by quadrupole ion filtering. Subsequently, immune complexes are dissociated by collision induced fragmentation and the ion signals of the "complex-released peptides," which in effect are the epitope peptides, are recorded in the time-of-flight analyzer of the mass spectrometer. Mixing of an antibody solution with a solution in which antigens or antigen-derived peptides are dissolved is, together with antigen proteolysis, the only required in-solution handling step. Simplicity of sample handling and speed of analysis together with very low sample consumption makes ITEM faster and easier to perform than other experimental epitope mapping methods. Graphical Abstract ᅟ.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-hnRNP-A2/B1 antibody, Mouse monoclonal, clone DP3B3, purified from hybridoma cell culture
Sigma-Aldrich
Diethylumbelliferyl phosphate, ≥98% (HPLC), oil