MilliporeSigma
  • Excitatory Modulation of the preBötzinger Complex Inspiratory Rhythm Generating Network by Endogenous Hydrogen Sulfide.
Excitatory Modulation of the preBötzinger Complex Inspiratory Rhythm Generating Network by Endogenous Hydrogen Sulfide.
Frontiers in physiology (2017-07-18)
Glauber S F da Silva, João P J Sabino, Vishaal Rajani, Tucaauê S Alvares, Silvia Pagliardini, Luiz G S Branco, Gregory D Funk
ABSTRACT
Hydrogen Sulfide (H
Sigma-Aldrich
N-Methyl-D-aspartic acid, ≥98% (TLC), solid