MilliporeSigma
  • Noncanonical Function of a Small-Molecular Virulence Factor Coronatine against Plant Immunity: An
Noncanonical Function of a Small-Molecular Virulence Factor Coronatine against Plant Immunity: An
ACS central science (2017-06-03)
Minoru Ueda, Syusuke Egoshi, Kosuke Dodo, Yasuhiro Ishimaru, Hiroyuki Yamakoshi, Takeshi Nakano, Yousuke Takaoka, Shinya Tsukiji, Mikiko Sodeoka
ABSTRACT
Coronatine (
Sigma-Aldrich
Coronatine, from Pseudomonas syringae pv Glycinea, >95% (HPLC)