MilliporeSigma
  • Quality and characteristics of fermented ginseng seed oil based on bacterial strain and extraction method.
Quality and characteristics of fermented ginseng seed oil based on bacterial strain and extraction method.
Journal of ginseng research (2017-07-14)
Myung-Hee Lee, Young-Kyoung Rhee, Sang-Yoon Choi, Chang-Won Cho, Hee-Do Hong, Kyung-Tack Kim
ABSTRACT
In this study, the fermentation of ginseng seeds was hypothesized to produce useful physiologically-active substances, similar to that observed for fermented ginseng root. Ginseng seed was fermented using The ginseng seed was fermented inoculating 1% of each strain on sterilized ginseng seeds and incubating the seeds at 30Ā°C for 24 h. Oil was extracted from the fermented ginseng seeds using compression extraction, solvent extraction, and supercritical fluid extraction. The color of the fermented ginseng seed oil did not differ greatly according to the fermentation or extraction method. The highest phenolic compound content recovered with the use of supercritical fluid extraction combined with fermentation using the
Sigma-Aldrich
Campesterol, ~65%