MilliporeSigma
  • Inhibition of Staphylococcus aureus in vitro by bacteriocinogenic Lactococcus lactis KTH0-1S isolated from Thai fermented shrimp (Kung-som) and safety evaluation.
Inhibition of Staphylococcus aureus in vitro by bacteriocinogenic Lactococcus lactis KTH0-1S isolated from Thai fermented shrimp (Kung-som) and safety evaluation.
Archives of microbiology (2017-01-07)
Sutanate Saelao, Suppasil Maneerat, Sireewan Kaewsuwan, Hanitra Rabesona, Yvan Choiset, Thomas Haertlé, Jean-Marc Chobert
ABSTRACT
Lactococcus lactis KTH0-1S isolated from Thai traditional fermented shrimp (Kung-som) is able to produce heat-stable bacteriocin and inhibits food spoilage bacteria and food-borne pathogens. The inhibitory effect of bacteriocin remained intact after treatment with different pHs and after heating, but was sensitive to some proteolytic enzymes. Addition of bacteriocin KTH0-1S to Staphylococcus aureus cultures decreased viable cell counts by 2.8 log CFU/ml, demonstrating a bactericidal mode of action. Furthermore, the growth of S. aureus decreased significantly after 12-h co-cultivation with bacteriocinogenic strain. The molecular mass of bacteriocin KTH0-1S was found to be 3.346 kDa after ammonium sulfate precipitation, reversed phase (C
Product Description
Supelco
L-Histidine hydrochloride solution, 100 mM amino acid in 0.1 M HCl, analytical standard