MilliporeSigma
  • Second-order nonlinear polarizability of ferrocene-BODIPY donor-acceptor adducts. Quantifying charge redistribution in the excited state.
Second-order nonlinear polarizability of ferrocene-BODIPY donor-acceptor adducts. Quantifying charge redistribution in the excited state.
Dalton transactions (Cambridge, England : 2003) (2017-01-04)
Navdeep Kaur, Nick Van Steerteghem, Priya Singla, Paramjit Kaur, Koen Clays, Kamaljit Singh
ABSTRACT
A series of dyads and triads using ferrocene (Fc) as the donor and 4,4-difluoro-4-bora-3a,4a-diaza-s-indacene (BODIPY) as the acceptor, linked either directly or through an N-phenylmethanimine or ethynylbenzene linker have been synthesized. While the former (directly linked) dyads were prepared through acid catalyzed condensation of pyrrole with ferrocenecarboxaldehye or 1,1'-ferrocenedicarboxaldehyde followed by oxidation with 2,3-dichloro-5,6-dicyano-1,4-benzoquinone (DDQ), the latter two sets (imine and alkyne linked) of dyads were obtained through Schiff base condensation or Sonogashira coupling reactions, respectively. The compounds were fully characterized with spectroscopic data and single crystal X-ray analysis in one case. The peaks corresponding to the Fe(ii)/Fe(iii) redox couple at 0.33 to 0.38 V showed a varying degree of positive anodic shift, which reflected the strong electron withdrawing effect of the corresponding BODIPY unit. The first hyperpolarisability, β, was measured in chloroform using the femtosecond hyper-Rayleigh scattering (HRS) method at 1300 nm. Interestingly, from the β
Sigma-Aldrich
1,3-Propylene sulfite, 99%