MilliporeSigma
  • Tlx3 Function in the Dorsal Root Ganglion is Pivotal to Itch and Pain Sensations.
Tlx3 Function in the Dorsal Root Ganglion is Pivotal to Itch and Pain Sensations.
Frontiers in molecular neuroscience (2017-07-14)
Chengcheng Huang, Fumin Lu, Ping Li, Cheng Cao, Zijing Liu
ABSTRACT
Itch, a sensation eliciting a desire to scratch, is distinct from but not completely independent of pain. Inspiring achievements have been made in the characterization of itch-related receptors and neurotransmitters, but the molecular mechanisms controlling the development of pruriceptors remain poorly understood. Here, our RNAseq and
Sigma-Aldrich
PAR-2 (1-6) amide (mouse, rat) trifluoroacetate salt, ≥97% (HPLC)
Sigma-Aldrich
Gastrin Releasing Peptide human, ≥97% (HPLC)