MilliporeSigma
One-Pot Green Synthesis of Ag-Decorated SnO
Nanoscale research letters (2017-07-05)
Min Hu, Zhenwei Zhang, Chenkun Luo, Xiuqing Qiao
ABSTRACT
In this paper, hierarchical Ag-decorated SnO
Sigma-Aldrich
Potassium borohydride, purum, ≥97.0% (RT)