MilliporeSigma
  • Down-regulation of the mitochondrial aspartate-glutamate carrier isoform 1 AGC1 inhibits proliferation and N-acetylaspartate synthesis in Neuro2A cells.
Down-regulation of the mitochondrial aspartate-glutamate carrier isoform 1 AGC1 inhibits proliferation and N-acetylaspartate synthesis in Neuro2A cells.
Biochimica et biophysica acta (2017-02-27)
Emanuela Profilo, Luis Emiliano Peña-Altamira, Mariangela Corricelli, Alessandra Castegna, Alberto Danese, Gennaro Agrimi, Sabrina Petralla, Giulia Giannuzzi, Vito Porcelli, Luigi Sbano, Carlo Viscomi, Francesca Massenzio, Erika Mariana Palmieri, Carlotta Giorgi, Giuseppe Fiermonte, Marco Virgili, Luigi Palmieri, Massimo Zeviani, Paolo Pinton, Barbara Monti, Ferdinando Palmieri, Francesco Massimo Lasorsa
ABSTRACT
The mitochondrial aspartate-glutamate carrier isoform 1 (AGC1) catalyzes a Ca
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose, sodium pyruvate, and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Tetramethylrhodamine methyl ester perchlorate, ≥95%
Sigma-Aldrich
Minimum Essential Medium Eagle, With Earle′s salts, non-essential amino acids and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture