MilliporeSigma
  • Three-dimensional structural analysis reveals a Cdk5-mediated kinase cascade regulating hepatic biliary network branching in zebrafish.
Three-dimensional structural analysis reveals a Cdk5-mediated kinase cascade regulating hepatic biliary network branching in zebrafish.
Development (Cambridge, England) (2017-07-20)
Manali Dimri, Cassandra Bilogan, Lain X Pierce, Gregory Naegele, Amit Vasanji, Isabel Gibson, Allyson McClendon, Kevin Tae, Takuya F Sakaguchi
ABSTRACT
The intrahepatic biliary network is a highly branched three-dimensional network lined by biliary epithelial cells, but how its branching patterns are precisely established is not clear. We designed a new computer-based algorithm that quantitatively computes the structural differences of the three-dimensional networks. Utilizing the algorithm, we showed that inhibition of Cyclin-dependent kinase 5 (Cdk5) led to reduced branching in the intrahepatic biliary network in zebrafish. Further, we identified a previously unappreciated downstream kinase cascade regulated by Cdk5. Pharmacological manipulations of this downstream kinase cascade produced a crowded branching defect in the intrahepatic biliary network and influenced actin dynamics in biliary epithelial cells. We generated larvae carrying a mutation in
Sigma-Aldrich
Cucurbitacin E, ≥95% (HPLC)
Sigma-Aldrich
Olomoucine, ≥98% (HPLC)
Sigma-Aldrich
IPA-3, ≥95% (HPLC), powder
Sigma-Aldrich
LIM Kinase Inhibitor I, LIMKi 3, The LIM Kinase Inhibitor I, LIMKi 3 controls the biological activity of LIM Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.