MilliporeSigma
  • D816V mutation in the KIT gene activation loop has greater cell-proliferative and anti-apoptotic ability than N822K mutation in core-binding factor acute myeloid leukemia.
D816V mutation in the KIT gene activation loop has greater cell-proliferative and anti-apoptotic ability than N822K mutation in core-binding factor acute myeloid leukemia.
Experimental hematology (2017-05-17)
Ikuko Omori, Hiroki Yamaguchi, Koichi Miyake, Noriko Miyake, Tomoaki Kitano, Koiti Inokuchi
ABSTRACT
In core-binding factor acute myeloid leukemia (CBF-AML), there have been conflicting reports regarding the status as an unfavorable prognostic factor of mutation in the KIT gene, the significance of which remains unclear. We previously reported that prognoses differ between the KIT D816V and N822K mutations. In the present study, we compared in vitro the cell-proliferative and anti-apoptotic ability of D816V and N822K. We transduced these KIT mutations into the interleukin-3-dependent cell line TF-1 (TF-1 KIT
Sigma-Aldrich
Midostaurin hydrate, ≥98% (HPLC), solid
Sigma-Aldrich
Imatinib
Sigma-Aldrich
Dasatinib