MilliporeSigma
  • Fitness and inheritance of metaflumizone resistance in Plutella xylostella.
Fitness and inheritance of metaflumizone resistance in Plutella xylostella.
Pesticide biochemistry and physiology (2017-06-10)
Jun Shen, Dongyang Li, Shuzhen Zhang, Xun Zhu, Hu Wan, Jianhong Li
ABSTRACT
The diamondback moth, Plutella xylostella (L.) has developed resistance to many types of insecticides in the field. To study inheritance and fitness cost of metaflumizone resistance, a susceptible strain of diamondback moth was continuously selected with metaflumizone during 37 generations under laboratory conditions. The resistance to metaflumizone was at a high level (resistance ratios from 250.37 to 1450.47-fold). We investigated a metaflumizone resistance strain (G
Product Description
Supelco
Metaflumizone, PESTANAL®, analytical standard