MilliporeSigma
  • Insulin resistance and normal thyroid hormone levels: prospective study and metabolomic analysis.
Insulin resistance and normal thyroid hormone levels: prospective study and metabolomic analysis.
American journal of physiology. Endocrinology and metabolism (2017-03-02)
Ele Ferrannini, Giorgio Iervasi, Jeff Cobb, Rudina Ndreu, Monica Nannipieri
ABSTRACT
While hyperthyroidism and hypothyroidism cause dysglycemia, the relationship between thyroid hormone levels within the normal range and insulin resistance (IR) is unclear. In 940 participants with strictly normal serum concentrations of free triiodothyronine (fT