MilliporeSigma
  • The effects of ingestion of hormonal host factors on the longevity and insecticide resistance phenotype of the major malaria vector Anopheles arabiensis (Diptera: Culicidae).
The effects of ingestion of hormonal host factors on the longevity and insecticide resistance phenotype of the major malaria vector Anopheles arabiensis (Diptera: Culicidae).
PloS one (2017-07-13)
Shüné V Oliver, Basil D Brooke
ABSTRACT
Exogenous vertebrate-derived factors circulating in the blood have the capacity to modulate the biology of haematophagous insects. These include insulin, insulin growth factor 1 (IGF) and transforming growth factor β1 (TGFβ). The effects of the consumption of these three proteins were examined on laboratory strains of Anopheles arabiensis. SENN, an insecticide susceptible strain and SENN DDT, a resistant strain selected from SENN, were fed with host factor-supplemented sucrose. Adult longevity was measured and insecticide resistance phenotype over time was assessed by WHO bioassay. Detoxification and oxidative stress defence enzyme activity was assessed calorimetrically. Insulin supplementation augmented insecticide resistance in young adult mosquitoes. This effect was due to the hormonal nature of the protein, as heat-denatured insulin did not elicit the same response. In contrast, IGF and TGFβ consumption generally reduced the expression of insecticide resistance. Insulin ingestion significantly reduced longevity in the insecticide susceptible strain. IGF elicited the same response in the susceptible strain, while TGF consumption had no effect on either strain. Consumption of all factors significantly decreased Glutathione S-transferase activity and increased cytochrome P450 and superoxide dismutase activity. This suggests that the altered detoxification phenotype is mediated primarily by cytochrome P450 activity, which would result in an increase in oxidative stress. The increased superoxide dismutase activity suggests that this enzyme class alleviates the oxidative stress as opposed to glutathione-based redox systems. Oxidative stress responses play a crucial role in insecticide resistance and longevity. These data show that ingested hormonal factors can affect mosquito longevity and insecticide susceptibility, both of which are important characteristics in terms of malaria transmission and control.
Sigma-Aldrich
Insulin human, recombinant, expressed in yeast (proprietary host)
Sigma-Aldrich
SOD Assay Kit, sufficient for 500 tests
Sigma-Aldrich
BrdU (5-Bromo-2′-deoxyuridine), Used for the study of DNA synthesis where it is incorporated into DNA in place of thymidine, BrdU can be used in conjunction with anti-BrdU for immunocytochemical analysis of cell proliferation.