MilliporeSigma
  • Identification of FERM domain-containing protein 5 as a novel target of β-catenin/TCF7L2 complex.
Identification of FERM domain-containing protein 5 as a novel target of β-catenin/TCF7L2 complex.
Cancer science (2017-01-25)
Chi Zhu, Kiyoshi Yamaguchi, Tomoyuki Ohsugi, Yumi Terakado, Rei Noguchi, Tsuneo Ikenoue, Yoichi Furukawa
ABSTRACT
Deregulation of the canonical Wnt signaling pathway plays an important role in human tumorigenesis through the accumulation of β-catenin and subsequent transactivation of TCF7L2. Although some of the consequences associated with the accumulated β-catenin have been clarified, the comprehensive effect of activated β-catenin/TCF7L2 transcriptional complex on tumorigenesis remains to be elucidated. To understand the precise molecular mechanisms underlying colorectal cancer, we searched for genes regulated by the complex in colorectal tumors. We performed expression profile analysis of HCT116 and SW480 colon cancer cells treated with β-catenin siRNAs, and ChIP-sequencing using anti-TCF7L2 antibody. Combination of these data with public microarray data of LS174 cells with a dominant-negative form of TCF7L2 identified a total of 11 candidate genes. In this paper, we focused on FERM domain-containing protein 5 (FRMD5), and confirmed that it is regulated by both β-catenin and TCF7L2. An additional reporter assay disclosed that a region in intron1 transcriptionally regulated the expression of FRMD5. ChIP assay also corroborated that TCF7L2 associates with this region. These data suggested that FRMD5 is a novel direct target of the β-catenin/TCF7L2 complex.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-TCF-4 Antibody, clone 6H5-3, clone 6H5-3, Upstate®, from mouse
Sigma-Aldrich
Anti-FRMD5 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution