MilliporeSigma
  • Effects of arachidonic acid on FFA4 receptor: Signaling, phosphorylation and internalization.
Effects of arachidonic acid on FFA4 receptor: Signaling, phosphorylation and internalization.
Prostaglandins, leukotrienes, and essential fatty acids (2017-02-27)
S Villegas-Comonfort, Y Takei, G Tsujimoto, A Hirasawa, J A García-Sáinz
ABSTRACT
Arachidonic acid increased intracellular calcium, in cells expressing green fluorescent protein-tagged human FFA4 receptors, with an EC
Sigma-Aldrich
AH-7614, ≥98% (HPLC)