MilliporeSigma
  • FYCO1 and autophagy control the integrity of the haploid male germ cell-specific RNP granules.
FYCO1 and autophagy control the integrity of the haploid male germ cell-specific RNP granules.
Autophagy (2016-12-09)
Matteo Da Ros, Tiina Lehtiniemi, Opeyemi Olotu, Daniel Fischer, Fu-Ping Zhang, Helena Vihinen, Eija Jokitalo, Anu Sironen, Jorma Toppari, Noora Kotaja
ABSTRACT
Ribonucleoprotein (RNP) granules play a major role in compartmentalizing cytoplasmic RNA regulation. Haploid round spermatids that have exceptionally diverse transcriptomes are characterized by a unique germ cell-specific RNP granule, the chromatoid body (CB). The CB shares many characteristics with somatic RNP granules but also has germline-specific features. The CB appears to be a central structure in PIWI-interacting RNA (piRNA)-targeted RNA regulation. Here, we identified a novel CB component, FYCO1, which is involved in the intracellular transport of autophagic vesicles in somatic cells. We demonstrated that the CB is associated with autophagic activity. Induction of autophagy leads to the recruitment of lysosomal vesicles onto the CB in a FYCO1-dependent manner as demonstrated by the analysis of a germ cell-specific Fyco1 conditional knockout mouse model. Furthermore, in the absence of FYCO1, the integrity of the CB was affected and the CB was fragmented. Our results suggest that RNP granule homeostasis is regulated by FYCO1-mediated autophagy.
Sigma-Aldrich
Anti-RUVBL1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
Sigma-Aldrich
Wortmannin, from Penicillium funiculosum, ≥98% (HPLC and TLC)
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Roche
cOmplete Protease Inhibitor Cocktail, Tablets provided in EASYpacks
Sigma-Aldrich
TWEEN® 20, viscous liquid, suitable for cell culture
Sigma-Aldrich
Anti-FYCO1 antibody produced in mouse, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)