MilliporeSigma
  • Deep Sequencing of the Human Retinae Reveals the Expression of Odorant Receptors.
Deep Sequencing of the Human Retinae Reveals the Expression of Odorant Receptors.
Frontiers in cellular neuroscience (2017-02-09)
Nikolina Jovancevic, Kirsten A Wunderlich, Claudia Haering, Caroline Flegel, Désirée Maßberg, Markus Weinrich, Lea Weber, Lars Tebbe, Anselm Kampik, Günter Gisselmann, Uwe Wolfrum, Hanns Hatt, Lian Gelis
ABSTRACT
Several studies have demonstrated that the expression of odorant receptors (ORs) occurs in various tissues. These findings have served as a basis for functional studies that demonstrate the potential of ORs as drug targets for a clinical application. To the best of our knowledge, this report describes the first evaluation of the mRNA expression of ORs and the localization of OR proteins in the human retina that set a stage for subsequent functional analyses. RNA-Sequencing datasets of three individual neural retinae were generated using Next-generation sequencing and were compared to previously published but reanalyzed datasets of the peripheral and the macular human retina and to reference tissues. The protein localization of several ORs was investigated by immunohistochemistry. The transcriptome analyses detected an average of 14 OR transcripts in the neural retina, of which
Sigma-Aldrich
Anti-OR10AD1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-OR5P3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-OR2W3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution