MilliporeSigma
  • BMP-induced reprogramming of the neural retina into retinal pigment epithelium requires Wnt signalling.
BMP-induced reprogramming of the neural retina into retinal pigment epithelium requires Wnt signalling.
Biology open (2017-05-27)
Jörg Steinfeld, Ichie Steinfeld, Alexander Bausch, Nicola Coronato, Meggi-Lee Hampel, Heike Depner, Paul G Layer, Astrid Vogel-Höpker
ABSTRACT
In vertebrates, the retinal pigment epithelium (RPE) and photoreceptors of the neural retina (NR) comprise a functional unit required for vision. During vertebrate eye development, a conversion of the RPE into NR can be induced by growth factors
Sigma-Aldrich
Anti-MITF antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution