MilliporeSigma
  • Determination of Cu/Zn and Fe in human serum of patients with sickle cell anemia using radiation synchrotron.
Determination of Cu/Zn and Fe in human serum of patients with sickle cell anemia using radiation synchrotron.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine (2012-03-01)
C G L Canellas, S M F Carvalho, M J Anjos, R T Lopes
ABSTRACT
In this work we analyzed serum samples from patients with Sickle Cell Anemia (SCA) using Total Reflection X-Ray Fluorescence using Synchrotron Radiation (SRTXRF). The SRTXRF measurements were performed at the X-Ray Fluorescence Beamline at the Brazilian National Synchrotron Light Laboratory (LNLS). We studied forty-three patients aged 18-50 suffering from SCA and sixty healthy volunteers aged 18-60. It was possible to determine the concentrations of the following elements: P, S, Cl, K, Ca, Fe, Cu, Zn, Br and Rb. Moreover, there are evidences of an association among Fe, Cu, Zn and Cu/Zn in the SCA pathogenesis process. The concentrations of Fe and Cu in the serum samples of patients with SCA were larger, 120% and 20%, respectively, when compared with the CG. The serum level Cu/Zn ratio was significantly higher (60%) in the serum samples from patients suffering from SCA than from the CG. Therefore, the Cu/Zn ratio can be used as an adjuvant index in enhancement for diagnosis of SCA.
Product Description
Supelco
Gallium Standard for ICP, TraceCERT®, 1 g/L Ga in nitric acid (nominal concentration)