MilliporeSigma
  • High frequency of LMAN1 abnormalities in colorectal tumors with microsatellite instability.
High frequency of LMAN1 abnormalities in colorectal tumors with microsatellite instability.
Cancer research (2009-01-02)
Nina Roeckel, Stefan M Woerner, Matthias Kloor, Yan-Ping Yuan, Georgios Patsos, Roland Gromes, Juergen Kopitz, Johannes Gebert
ABSTRACT
Glycosyl epitopes have been identified as tumor-specific markers in colorectal tumors and various lines of evidence indicate the significance of altered synthesis, transport, and secretion of glycoproteins in tumorigenesis. However, aberrant glycosylation has been largely ignored in microsatellite unstable (MSI-H) colorectal tumors. Therefore, we analyzed mutation frequencies of genes of the cellular glycosylation machinery in MSI-H tumors, focusing on frameshift mutations in coding MNRs (cMNRs). Among 28 candidate genes, LMAN1/ERGIC53, a mannose-specific lectin mediating endoplasmatic reticulum (ER)-to-Golgi transit of glycosylated proteins, showed high mutation frequency in MSI-H colorectal cancer cell lines (52%; 12 of 23), carcinomas (45%; 72 of 161), and adenomas (40%; 8 of 20). Biallelic mutations were observed in 17% (4 of 23) of MSI-H colorectal cancer cell lines. LMAN1 was found to be transcribed but truncated protein remained undetectable in these LMAN1-mutant cell lines. Immunohistochemical and molecular analysis of LMAN1-mutated carcinomas and adenomas revealed regional loss of LMAN1 expression due to biallelic LMAN1 cMNR frameshift mutations. In LMAN1-deficient colorectal cancer cell lines, secretion of the LMAN1 client protein alpha-1-antitrypsin (A1AT), an inhibitor of angiogenesis and tumor growth, was significantly impaired but could be restored upon LMAN1 re-expression. These results suggest that LMAN1 mutational inactivation is a frequent and early event potentially contributing to MSI-H tumorigenesis.