MilliporeSigma
  • Cysteine and glycine-rich protein 2 (CSRP2) transcript levels correlate with leukemia relapse and leukemia-free survival in adults with B-cell acute lymphoblastic leukemia and normal cytogenetics.
Cysteine and glycine-rich protein 2 (CSRP2) transcript levels correlate with leukemia relapse and leukemia-free survival in adults with B-cell acute lymphoblastic leukemia and normal cytogenetics.
Oncotarget (2017-04-19)
Shu-Juan Wang, Ping-Zhang Wang, Robert Peter Gale, Ya-Zhen Qin, Yan-Rong Liu, Yue-Yun Lai, Hao Jiang, Qian Jiang, Xiao-Hui Zhang, Bin Jiang, Lan-Ping Xu, Xiao-Jun Huang, Kai-Yan Liu, Guo-Rui Ruan
ABSTRACT
Relapse is the major cause of treatment-failure in adults with B-cell acute lymphoblastic leukemia (ALL) achieving complete remission after induction chemotherapy. Greater precision identifying persons likely to relapse is important. We did bio-informatics analyses of transcriptomic data to identify mRNA transcripts aberrantly-expressed in B-cell ALL. We selected 9 candidate genes for validation 7 of which proved significantly-associated with B-cell ALL. We next focused on function and clinical correlations of the cysteine and glycine-rich protein 2 (CSRP2). Quantitative real-time polymerase chain reaction (RT-qPCR) was used to examine gene transcript levels in bone marrow samples from 236 adults with B-cell ALL compared with samples from normals. CSRP2 was over-expressed in 228 out of 236 adults (97%) with newly-diagnosed B-cell ALL. A prognostic value was assessed in 168 subjects. In subjects with normal cytogenetics those with high CSRP2 transcript levels had a higher 5-year cumulative incidence of relapse (CIR) and worse relapse-free survival (RFS) compared with subjects with low transcript levels (56% [95% confidence interval, 53, 59%] vs. 19% [18, 20%]; P = 0.011 and 41% [17, 65%] vs. 80% [66-95%]; P = 0.007). In multivariate analyses a high CSRP2 transcript level was independently-associated with CIR (HR = 5.32 [1.64-17.28]; P = 0.005) and RFS (HR = 5.56 [1.87, 16.53]; P = 0.002). Functional analyses indicated CSRP2 promoted cell proliferation, cell-cycle progression, in vitro colony formation and cell migration ability. Abnormal CSRP2 expression was associated with resistance to chemotherapy; sensitivity was restored by down-regulating CSRP2 expression.
Sigma-Aldrich
MISSION® esiRNA, targeting human CSRP2