MilliporeSigma
  • Histone deacetylase 10 structure and molecular function as a polyamine deacetylase.
Histone deacetylase 10 structure and molecular function as a polyamine deacetylase.
Nature communications (2017-05-19)
Yang Hai, Stephen A Shinsky, Nicholas J Porter, David W Christianson
ABSTRACT
Cationic polyamines such as spermidine and spermine are critical in all forms of life, as they regulate the function of biological macromolecules. Intracellular polyamine metabolism is regulated by reversible acetylation and dysregulated polyamine metabolism is associated with neoplastic diseases such as colon cancer, prostate cancer and neuroblastoma. Here we report that histone deacetylase 10 (HDAC10) is a robust polyamine deacetylase, using recombinant enzymes from Homo sapiens (human) and Danio rerio (zebrafish). The 2.85 Å-resolution crystal structure of zebrafish HDAC10 complexed with a transition-state analogue inhibitor reveals that a glutamate gatekeeper and a sterically constricted active site confer specificity for N
Sigma-Aldrich
Maltose solution, BioReagent, ~20% in H2O
Sigma-Aldrich
N-butylacetamide, AldrichCPR
Sigma-Aldrich
N-(4-Aminobutyl)acetamide