MilliporeSigma
  • Characterization of the Anti-PD-1 Antibody REGN2810 and Its Antitumor Activity in Human
Characterization of the Anti-PD-1 Antibody REGN2810 and Its Antitumor Activity in Human
Molecular cancer therapeutics (2017-03-08)
Elena Burova, Aynur Hermann, Janelle Waite, Terra Potocky, Venus Lai, Seongwon Hong, Matt Liu, Omaira Allbritton, Amy Woodruff, Qi Wu, Amanda D'Orvilliers, Elena Garnova, Ashique Rafique, William Poueymirou, Joel Martin, Tammy Huang, Dimitris Skokos, Joel Kantrowitz, Jon Popke, Markus Mohrs, Douglas MacDonald, Ella Ioffe, William Olson, Israel Lowy, Andrew Murphy, Gavin Thurston
ABSTRACT
The Programmed Death-1 (PD-1) receptor delivers inhibitory checkpoint signals to activated T cells upon binding to its ligands PD-L1 and PD-L2 expressed on antigen-presenting cells and cancer cells, resulting in suppression of T-cell effector function and tumor immune evasion. Clinical antibodies blocking the interaction between PD-1 and PD-L1 restore the cytotoxic function of tumor antigen-specific T cells, yielding durable objective responses in multiple cancers. This report describes the preclinical characterization of REGN2810, a fully human hinge-stabilized IgG4(S228P) high-affinity anti-PD-1 antibody that potently blocks PD-1 interactions with PD-L1 and PD-L2. REGN2810 was characterized in a series of binding, blocking, and functional cell-based assays, and preclinical
Sigma-Aldrich
Human PDCD1 / Programmed Cell Death Protein 1 ELISA Kit