MilliporeSigma
  • Structure-activity studies on polymyxin derivatives carrying three positive charges only reveal a new class of compounds with strong antibacterial activity.
Structure-activity studies on polymyxin derivatives carrying three positive charges only reveal a new class of compounds with strong antibacterial activity.
Peptides (2017-03-17)
Martti Vaara, Timo Vaara, Jonathan M Tyrrell
ABSTRACT
Recent years have brought in an increased interest to develop improved polymyxins. The currently used polymyxins, i.e. polymyxin B and colistin (polymyxin E) are pentacationic lipopeptides that possess a cyclic heptapeptide part with three positive charges, a linear "panhandle" part with two positive charges, and a fatty acyl tail. Unfortunately, their clinical use is shadowed by their notable nephrotoxicity. We have previously developed a polymyxin derivative NAB739 which lacks the positive charges in the linear part. This derivative is better tolerated than polymyxin B in cynomolgus monkeys and is, in contrast to polymyxin B, excreted into urine in monkeys and rats. Here we have conducted further structure-activity relationship (SAR) studies on 17 derivatives with three positive charges only. We discovered a remarkably antibacterial class, as exemplified by NAB815, that carries two positive charges only in the cyclic part.
Sigma-Aldrich
Polymyxin B sulfate, meets USP testing specifications