MilliporeSigma
  • Interactions between Plant Metabolites Affect Herbivores: A Study with Pyrrolizidine Alkaloids and Chlorogenic Acid.
Interactions between Plant Metabolites Affect Herbivores: A Study with Pyrrolizidine Alkaloids and Chlorogenic Acid.
Frontiers in plant science (2017-06-15)
Xiaojie Liu, Klaas Vrieling, Peter G L Klinkhamer
ABSTRACT
The high structural diversity of plant metabolites suggests that interactions among them should be common. We investigated the effects of single metabolites and combinations of plant metabolites on insect herbivores. In particular we studied the interacting effects of pyrrolizidine alkaloid (PAs), and chlorogenic acid (CGA), on a generalist herbivore,
Product Description
Supelco
Abamectin, PESTANAL®, analytical standard
Sigma-Aldrich
3-Chloroperbenzoic acid, ≤77%