MilliporeSigma
Anti-allergic Hydroxy Fatty Acids from
Frontiers in pharmacology (2017-07-05)
Michal Korinek, Yi-Hong Tsai, Mohamed El-Shazly, Kuei-Hung Lai, Anders Backlund, Shou-Fang Wu, Wan-Chun Lai, Tung-Ying Wu, Shu-Li Chen, Yang-Chang Wu, Yuan-Bin Cheng, Tsong-Long Hwang, Bing-Hung Chen, Fang-Rong Chang
ABSTRACT
Increasing prevalence of allergic diseases with an inadequate variety of treatment drives forward search for new alternative drugs. Fatty acids, abundant in nature, are regarded as important bioactive compounds and powerful nutrients playing an important role in lipid homeostasis and inflammation. Phytochemical study on
Sigma-Aldrich
cis-Vaccenic acid, ≥97% (capillary GC), oil
Sigma-Aldrich
2OHOA, ≥98% (HPLC)
Sigma-Aldrich
DL-α-Hydroxystearic acid, ≥99%
Sigma-Aldrich
Methyl 12-oxooctadecanoate, 98%
Sigma-Aldrich
9(S)-HODE, ≥98% (HPLC)