MilliporeSigma
  • Peroxotantalate-Based Ionic Liquid Catalyzed Epoxidation of Allylic Alcohols with Hydrogen Peroxide.
Peroxotantalate-Based Ionic Liquid Catalyzed Epoxidation of Allylic Alcohols with Hydrogen Peroxide.
Chemistry (Weinheim an der Bergstrasse, Germany) (2017-04-06)
Wenbao Ma, Chen Chen, Kang Kong, Qifeng Dong, Kun Li, Mingming Yuan, Difan Li, Zhenshan Hou
ABSTRACT
The efficient and environmentally benign epoxidation of allylic alcohols has been attained by using new kinds of monomeric peroxotantalate anion-functionalized ionic liquids (ILs=[P
Sigma-Aldrich
1,3-Propylene sulfite, 99%
Sigma-Aldrich
Tetrabutylphosphonium chloride, 96%