MilliporeSigma
  • Synergic effects of 9,10-phenanthrenequinone and cadmium on pro-inflammatory responses in airway epithelial cells.
Synergic effects of 9,10-phenanthrenequinone and cadmium on pro-inflammatory responses in airway epithelial cells.
Environmental toxicology and pharmacology (2017-05-10)
Akiko Honda, Pratiti Home Chowdhury, Sho Ito, Hitoshi Okano, Toshinori Onishi, Yusuke Kawaryu, Kayo Ueda, Hirohisa Takano
ABSTRACT
We investigated the synergic effects of components of particulate matter with aerodynamic diameters ≤2.5μm (PM2.5) on airway inflammation. Co-exposure to cadmium (Cd) and 9,10-phenanthrenequinone (9,10-PQ) additively/synergistically increased pro-inflammatory responses in airway epithelial cells, whereas co-exposure to Cd and phenanthrene resulted in no acceleration. These results suggest that the combination of metal and a quinone derivative can contribute to the exacerbation of respiratory diseases by PM2.5.
Sigma-Aldrich
Cadmium sulfate hydrate, ≥99.995% trace metals basis