MilliporeSigma
  • Is there an association between enhanced choline and β-catenin pathway in breast cancer? A pilot study by MR Spectroscopy and ELISA.
Is there an association between enhanced choline and β-catenin pathway in breast cancer? A pilot study by MR Spectroscopy and ELISA.
Scientific reports (2017-05-24)
Khushbu Agarwal, Gururao Hariprasad, Komal Rani, Uma Sharma, Sandeep R Mathur, Vurthaluru Seenu, Rajinder Parshad, Naranamangalam R Jagannathan
ABSTRACT
Total choline (tCho) was documented as a biomarker for breast cancer diagnosis by in vivo MRS. To understand the molecular mechanisms behind elevated tCho in breast cancer, an association of tCho with β-catenin and cyclin D1 was evaluated. Hundred fractions from 20 malignant, 10 benign and 20 non-involved breast tissues were isolated. Cytosolic and nuclear expressions of β-catenin and cyclin D1 were estimated using ELISA. Higher tCho was seen in malignant compared to benign tissues. Malignant tissues showed higher cytosolic and nuclear β-catenin expressions than benign and non-involved tissues. Within malignant tissues, β-catenin and cyclin D1 expressions were higher in the nucleus than cytosol. Cyclin D1 expression was higher in the cytosolic fractions of benign and non-involved than malignant tissues. Furthermore, in malignant tissues, tCho showed a positive correlation with the cytosolic and nuclear expression of β-catenin and cyclin D1 and also a correlation between nuclear expressions of both these proteins was seen. Higher cytosolic β-catenin expression was seen in progesterone receptor negative than positive patients. Results provide an evidence of correlation between non-invasive biomarker, tCho and the Wnt/β-catenin pathway. The findings explain the molecular mechanism of tCho elevation which may facilitate exploration of additional therapeutic targets for breast cancer.
Sigma-Aldrich
ANTI-HIST1H2BL (N-TERM) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution