MilliporeSigma
  • Optimisation of combi-lipases from Aspergillus niger for the synergistic and efficient hydrolysis of soybean oil.
Optimisation of combi-lipases from Aspergillus niger for the synergistic and efficient hydrolysis of soybean oil.
Animal science journal = Nihon chikusan Gakkaiho (2016-10-06)
Hanzhen Qiao, Fei Zhang, Wutai Guan, Jianjun Zuo, Dingyuan Feng
ABSTRACT
The enzymatic properties of four lipases (A, B, C and D) from different strains of Aspergillus niger, were investigated, and a 3-factor mixture design and triangular surface analysis were performed to screen the optimal combi-lipase by observing synergistic effects. Lipases B and D differed in optimal pH, temperature and substrate specificity. A combi-lipase with 31.2% lipase B and 68.8% lipase D (w/w, equal to units of 30.36% and 69.64%) exhibited optimal hydrolytic activity on soybean oil, which exceeded the sum of the combined activities of individual lipases (P < 0.05). Free fatty acid from the hydrolyzed soybean oil indicated that the synergistic effect of the combi-lipase resulted from the different fatty acid specificities of the two lipases. Overall, combi-lipase afforded an effective route for the application of lipase enzymes to animal feeds.
Sigma-Aldrich
4-Nitrophenyl octanoate, ≥90.0% (GC)
Sigma-Aldrich
4-Nitrophenyl palmitate, lipase substrate
Sigma-Aldrich
4-Nitrophenyl stearate, lipase substrate
Sigma-Aldrich
4-Nitrophenyl dodecanoate, ≥98.0% (GC)
Sigma-Aldrich
4-Nitrophenyl butyrate, ≥98%