MilliporeSigma
  • A rapid and effective method for screening, sequencing and reporter verification of engineered frameshift mutations in zebrafish.
A rapid and effective method for screening, sequencing and reporter verification of engineered frameshift mutations in zebrafish.
Disease models & mechanisms (2017-03-11)
Sergey V Prykhozhij, Shelby L Steele, Babak Razaghi, Jason N Berman
ABSTRACT
Clustered regularly interspaced palindromic repeats (CRISPR)/Cas-based adaptive immunity against pathogens in bacteria has been adapted for genome editing and applied in zebrafish (Danio rerio) to generate frameshift mutations in protein-coding genes. Although there are methods to detect, quantify and sequence CRISPR/Cas9-induced mutations, identifying mutations in F1 heterozygous fish remains challenging. Additionally, sequencing a mutation and assuming that it causes a frameshift does not prove causality because of possible alternative translation start sites and potential effects of mutations on splicing. This problem is compounded by the relatively few antibodies available for zebrafish proteins, limiting validation at the protein level. To address these issues, we developed a detailed protocol to screen F1 mutation carriers, and clone and sequence identified mutations. In order to verify that mutations actually cause frameshifts, we created a fluorescent reporter system that can detect frameshift efficiency based on the cloning of wild-type and mutant cDNA fragments and their expression levels. As proof of principle, we applied this strategy to three CRISPR/Cas9-induced mutations in pycr1a, chd7 and hace1 genes. An insertion of seven nucleotides in pycr1a resulted in the first reported observation of exon skipping by CRISPR/Cas9-induced mutations in zebrafish. However, of these three mutant genes, the fluorescent reporter revealed effective frameshifting exclusively in the case of a two-nucleotide deletion in chd7, suggesting activity of alternative translation sites in the other two mutants even though pycr1a exon-skipping deletion is likely to be deleterious. This article provides a protocol for characterizing frameshift mutations in zebrafish, and highlights the importance of checking mutations at the mRNA level and verifying their effects on translation by fluorescent reporters when antibody detection of protein loss is not possible.
Sigma-Aldrich
Glycogen from mussel, Mytilus genus, for DNA precipitations
Sigma-Aldrich
SYBR® Green I nucleic acid gel stain, 10,000 × in DMSO
Sigma-Aldrich
Agarose, low gelling temperature, BioReagent
Sigma-Aldrich
N,N,N′,N′-Tetramethylethylenediamine, BioReagent, suitable for electrophoresis, ≥99.0%