MilliporeSigma
  • Role of ammonium ion and transition metals in the formation of secondary organic aerosol and metallo-organic complex within fog processed ambient deliquescent submicron particles collected in central part of Indo-Gangetic Plain.
Role of ammonium ion and transition metals in the formation of secondary organic aerosol and metallo-organic complex within fog processed ambient deliquescent submicron particles collected in central part of Indo-Gangetic Plain.
Chemosphere (2017-05-10)
Dharmendra Kumar Singh, Tarun Gupta
ABSTRACT
In this study we observed the role of ammonium ion (NH
Product Description
Supelco
Multielement standard solution 5 for ICP, TraceCERT®, in 10% nitric acid