MilliporeSigma
  • SSRP1 Cooperates with PARP and XRCC1 to Facilitate Single-Strand DNA Break Repair by Chromatin Priming.
SSRP1 Cooperates with PARP and XRCC1 to Facilitate Single-Strand DNA Break Repair by Chromatin Priming.
Cancer research (2017-04-19)
Ying Gao, Changling Li, Leizhen Wei, Yaqun Teng, Satoshi Nakajima, Xiukai Chen, Jianquan Xu, Brittany Leger, Hongqiang Ma, Stephen T Spagnol, Yong Wan, Kris Noel Dahl, Yang Liu, Arthur S Levine, Li Lan
ABSTRACT
DNA single-strand breaks (SSB) are the most common form of DNA damage, requiring repair processes that to initiate must overcome chromatin barriers. The FACT complex comprised of the SSRP1 and SPT16 proteins is important for maintaining chromatin integrity, with SSRP1 acting as an histone H2A/H2B chaperone in chromatin disassembly during DNA transcription, replication, and repair. In this study, we show that SSRP1, but not SPT16, is critical for cell survival after ionizing radiation or methyl methanesulfonate-induced single-strand DNA damage. SSRP1 is recruited to SSB in a PARP-dependent manner and retained at DNA damage sites by N-terminal interactions with the DNA repair protein XRCC1. Mutational analyses showed how SSRP1 function is essential for chromatin decondensation and histone H2B exchange at sites of DNA strand breaks, which are both critical to prime chromatin for efficient SSB repair and cell survival. By establishing how SSRP1 facilitates SSB repair, our findings provide a mechanistic rationale to target SSRP1 as a general approach to selectively attack cancer cells. Cancer Res; 77(10); 2674-85. ©2017 AACR.
Sigma-Aldrich
MISSION® esiRNA, targeting human SSRP1
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Methyl methanesulfonate, 99%