MilliporeSigma
  • Preclinical anti-myeloma activity of EDO-S101, a new bendamustine-derived molecule with added HDACi activity, through potent DNA damage induction and impairment of DNA repair.
Preclinical anti-myeloma activity of EDO-S101, a new bendamustine-derived molecule with added HDACi activity, through potent DNA damage induction and impairment of DNA repair.
Journal of hematology & oncology (2017-06-22)
Ana-Alicia López-Iglesias, Ana B Herrero, Marta Chesi, Laura San-Segundo, Lorena González-Méndez, Susana Hernández-García, Irena Misiewicz-Krzeminska, Dalia Quwaider, Montserrat Martín-Sánchez, Daniel Primo, Teresa Paíno, P Leif Bergsagel, Thomas Mehrling, Marcos González-Díaz, Jesús F San-Miguel, María-Victoria Mateos, Norma C Gutiérrez, Mercedes Garayoa, Enrique M Ocio
ABSTRACT
Despite recent advances in the treatment of multiple myeloma (MM), the prognosis of most patients remains poor, and resistance to traditional and new drugs frequently occurs. EDO-S101 is a novel therapeutic agent conceived as the fusion of a histone deacetylase inhibitor radical to bendamustine, with the aim of potentiating its alkylating activity. The efficacy of EDO-S101 was evaluated in vitro, ex vivo and in vivo, alone, and in combination with standard anti-myeloma agents. The underlying mechanisms of action were also evaluated on MM cell lines, patient samples, and different murine models. EDO-S101 displayed potent activity in vitro in MM cell lines (IC These findings provide rationale for the clinical investigation of EDO-S101 in MM, either as a single agent or in combination with other anti-MM drugs, particularly proteasome inhibitors.
Sigma-Aldrich
Mirin, ≥98% (HPLC), powder
Sigma-Aldrich
Pomalidomide, ≥98% (HPLC)