MilliporeSigma
  • Targeted calcium influx boosts cytotoxic T lymphocyte function in the tumour microenvironment.
Targeted calcium influx boosts cytotoxic T lymphocyte function in the tumour microenvironment.
Nature communications (2017-05-16)
Kyun-Do Kim, Seyeon Bae, Tara Capece, Hristina Nedelkovska, Rafael G de Rubio, Alan V Smrcka, Chang-Duk Jun, Woojin Jung, Byeonghak Park, Tae-Il Kim, Minsoo Kim
ABSTRACT
Adoptive cell transfer utilizing tumour-targeting cytotoxic T lymphocytes (CTLs) is one of the most effective immunotherapies against haematological malignancies, but significant clinical success has not yet been achieved in solid tumours due in part to the strong immunosuppressive tumour microenvironment. Here, we show that suppression of CTL killing by CD4+CD25+Foxp3+ regulatory T cell (Treg) is in part mediated by TGFβ-induced inhibition of inositol trisphosphate (IP3) production, leading to a decrease in T cell receptor (TCR)-dependent intracellular Ca2+ response. Highly selective optical control of Ca2+ signalling in adoptively transferred CTLs enhances T cell activation and IFN-γ production in vitro, leading to a significant reduction in tumour growth in mice. Altogether, our findings indicate that the targeted optogenetic stimulation of intracellular Ca2+ signal allows for the remote control of cytotoxic effector functions of adoptively transferred T cells with outstanding spatial resolution by boosting T cell immune responses at the tumour sites.
Sigma-Aldrich
Anti-phospho-CD3ζ (pTyr142) antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution