MilliporeSigma
  • SERS Detection of Biomolecules by Highly Sensitive and Reproducible Raman-Enhancing Nanoparticle Array.
SERS Detection of Biomolecules by Highly Sensitive and Reproducible Raman-Enhancing Nanoparticle Array.
Nanoscale research letters (2017-05-13)
Tzu-Yi Chan, Ting-Yu Liu, Kuan-Syun Wang, Kun-Tong Tsai, Zhi-Xin Chen, Yu-Chi Chang, Yi-Qun Tseng, Chih-Hao Wang, Juen-Kai Wang, Yuh-Lin Wang
ABSTRACT
This paper describes the preparation of nanoarrays composed of silver nanoparticles (AgNPs: 20-50 nm) for use as surface-enhanced Raman scattering (SERS) substrates. The AgNPs were grown on porous anodic aluminum oxide (AAO) templates by electrochemical plating, and the inter-channel gap of AAO channels is between 10 and 20 nm. The size and interparticle gap of silver particles were adjusted in order to achieve optimal SERS signals and characterized by scanning electron microscopy, atomic force microscopy, and Raman spectroscopy. The fluctuation of SERS intensity is about 10-20% when measuring adenine solutions, showing a great reproducible SERS sensing. The nanoparticle arrays offer a large potential for practical applications as shown by the SERS-based quantitative detection and differentiation of adenine (A), thymine (T), cytosine (C), guanine (G), β-carotene, and malachite green. The respective detection limits are <1 ppb for adenine and <0.63 ppm for β-carotene and malachite green, respectively. Uniform and reproducible Raman enhancement enabled by Ag nanoparticle array embedded in anodic aluminum oxide differentiates and helps quantify DNA canonical nucleobases (adenine, thymine, cytosine, and guanine).
Product Description
Supelco
Malachite Green chloride, analytical standard