MilliporeSigma
Raptor regulates functional maturation of murine beta cells.
Nature communications (2017-06-10)
Qicheng Ni, Yanyun Gu, Yun Xie, Qinglei Yin, Hongli Zhang, Aifang Nie, Wenyi Li, Yanqiu Wang, Guang Ning, Weiqing Wang, Qidi Wang
ABSTRACT
Diabetes is associated with beta cell mass loss and islet dysfunctions. mTORC1 regulates beta cell survival, proliferation and function in physiological and pathological conditions, such as pregnancy and pancreatectomy. Here we show that deletion of Raptor, which is an essential component of mTORC1, in insulin-expressing cells promotes hypoinsulinemia and glucose intolerance. Raptor-deficient beta cells display reduced glucose responsiveness and exhibit a glucose metabolic profile resembling fetal beta cells. Knockout islets have decreased expression of key factors of functional maturation and upregulation of neonatal markers and beta cell disallowed genes, resulting in loss of functional maturity. Mechanistically, Raptor-deficient beta cells show reduced expression of DNA-methyltransferase 3a and altered patterns of DNA methylation at loci that are involved in the repression of disallowed genes. The present findings highlight a novel role of mTORC1 as a core mechanism governing postnatal beta cell maturation and physiologic beta cell mass during adulthood.
Sigma-Aldrich
Adenosine 5′-triphosphate (ATP) bioluminescent somatic cell assay kit, for cellular ATP determination