MilliporeSigma
  • New Wide Band Gap Donor for Efficient Fullerene-Free All-Small-Molecule Organic Solar Cells.
New Wide Band Gap Donor for Efficient Fullerene-Free All-Small-Molecule Organic Solar Cells.
Journal of the American Chemical Society (2017-01-14)
Liyan Yang, Shaoqing Zhang, Chang He, Jianqi Zhang, Huifeng Yao, Yang Yang, Yun Zhang, Wenchao Zhao, Jianhui Hou
ABSTRACT
A new organic small molecule, DRTB-T, that incorporates a two-dimensional trialkylthienyl-substituted benzodithiophene core building block was designed and synthesized. DRTB-T has a band gap (E
Sigma-Aldrich
1,1′-[4,8-Bis[5-(2-ethylhexyl)-2-thienyl]benzo[1,2-b:4,5-b′]dithiophene-2,6-diyl]bis[1,1,1-trimethylstannane]