MilliporeSigma
  • Maximization of monomeric C5 sugars from wheat bran by using mesoporous ordered silica catalysts.
Maximization of monomeric C5 sugars from wheat bran by using mesoporous ordered silica catalysts.
Bioresource technology (2017-04-30)
Nuria Sánchez-Bastardo, Esther Alonso
ABSTRACT
The hydrolysis process of a real fraction of arabinoxylans derived from wheat bran was studied. The influence of catalyst type and loading, reaction time and different metal cations were discussed in terms of the hydrolysis yield of arabinose and xylose oligomers as well as the formation of furfural as degradation product. A high yield of arabinoxylans into the corresponding monomeric sugars (96 and 94% from arabino- and xylo-oligosaccharides, respectively) was obtained at relatively high temperatures (180°C) and short reaction times (15min) with a catalyst loading of 4.8g of RuCl