MilliporeSigma
  • Activating transcription factor 3 promotes embryo attachment via up-regulation of leukemia inhibitory factor in vitro.
Activating transcription factor 3 promotes embryo attachment via up-regulation of leukemia inhibitory factor in vitro.
Reproductive biology and endocrinology : RB&E (2017-06-05)
Xi Cheng, Jingyu Liu, Huizhi Shan, Lihua Sun, Chenyang Huang, Qiang Yan, Ruiwei Jiang, Lijun Ding, Yue Jiang, Jianjun Zhou, Guijun Yan, Haixiang Sun
ABSTRACT
A receptive endometrium is essential for maternal-embryonic molecular communication during implantation. However, the specific molecular regulatory mechanisms of the endometrial capacity remain poorly understood. Here, we examined activating transcription factor 3 (ATF3) expression in human endometria and the functional effect of ATF3 on embryo attachment in vitro. Immunohistochemistry (IHC) was used to assess the ATF3 expression patterns in human endometria. Quantitative real-time PCR (qRT-PCR), western blotting and immunofluorescence (IF) studies were applied to explore ATF3 expression in Ishikawa cells upon estrogen (E2) and medroxyprogesterone acetate (MPA) treatment. qRT-PCR and western blotting were performed to inspect LIF (leukemia inhibitory factor) expression after enhancement or inhibition of ATF3, and a luciferase reporter assay and ChIP-PCR were used to confirm the regulatory mechanism of ATF3 to LIF. Endometrial epithelial capacity was assessed by an in vitro model of attachment of BeWo spheroids to Ishikawa cells. Western blotting was performed to compare the expression of ATF3 in endometrial samples of recurrent implantation failure (RIF) patients with that in samples from fertile women (FER) who had undergone no less than one successful embryo transplantation. ATF3 was located in human endometrial epithelial cells and stromal cells and was significantly induced by E2 and MPA in Ishikawa cells. Adenovirus-mediated overexpression of ATF3 in Ishikawa cells activated LIF promoter activity and enhanced its expression. Accordingly, the stimulation of BeWo spheroid adhesion promoted by ATF3 was inhibited by pretreatment with a specific antibody against LIF via the antibody-blocking assay. Moreover, ATF3 was aberrantly decreased in the endometria of RIF patients. Our findings suggest that ATF3 plays a significant role in regulating human endometrial receptivity and embryo attachment in vitro via up-regulation of leukemia inhibitory factor. Construction and management of the Nanjing multi-center biobank. No. 2013-081-01 . Registered 10 Dec. 2013.
Sigma-Aldrich
Monoclonal Anti-Leukemia Inhibitory Factor antibody produced in mouse, clone 9824, purified immunoglobulin, lyophilized powder
Sigma-Aldrich
Anti-ATF3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Phalloidin, Fluorescein Isothiocyanate Labeled, sequence Amanita phalloides(synthetic: peptide sequence)
Sigma-Aldrich
Mifepristone, ≥98%