MilliporeSigma
  • Alcohol Dependence Disrupts Amygdalar L-Type Voltage-Gated Calcium Channel Mechanisms.
Alcohol Dependence Disrupts Amygdalar L-Type Voltage-Gated Calcium Channel Mechanisms.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2017-04-02)
Florence P Varodayan, Giordano de Guglielmo, Marian L Logrip, Olivier George, Marisa Roberto
ABSTRACT
L-type voltage-gated calcium channels (LTCCs) are implicated in several psychiatric disorders that are comorbid with alcoholism and involve amygdala dysfunction. Within the amygdala, the central nucleus (CeA) is critical in acute alcohol's reinforcing actions, and its dysregulation in human alcoholics drives their negative emotional state and motivation to drink. Here we investigated the specific role of CeA LTCCs in the effects of acute alcohol at the molecular, cellular physiology, and behavioral levels, and their potential neuroadaptation in alcohol-dependent rats. Alcohol increases CeA activity (neuronal firing rates and GABA release) in naive rats by engaging LTCCs, and intra-CeA LTCC blockade reduces alcohol intake in nondependent rats. Alcohol dependence reduces CeA LTCC membrane abundance and disrupts this LTCC-based mechanism; instead, corticotropin-releasing factor type 1 receptors (CRF
Sigma-Aldrich
DNQX, ≥98% (TLC)
Sigma-Aldrich
Bisindolylmaleimide XI hydrochloride, ≥98% (TLC), solid
Sigma-Aldrich
Nifedipine, ≥98% (HPLC), powder