MilliporeSigma
LC-PDA-ESI-MS
Journal of mass spectrometry : JMS (2016-09-14)
Maja Friščić, Franz Bucar, Kroata Hazler Pilepić
ABSTRACT
Plants produce a great number of metabolites with potentially useful biological activities. Species from the genus Globularia (Plantaginaceae) are known as sources of different phenolic and iridoid compounds. Globularia alypum L. is a medicinal plant used as a healing agent in many Mediterranean countries. Similarities in phytochemical composition are often observed for related species. For Globularia spp., such findings were mostly based on identification of several isolated compounds from distinct species. To our knowledge, this is the first study that enables simultaneous comparison of phytochemical profiles from several members of the genus Globularia. Liquid chromatography-photodiode array detection-electrospray ionization-tandem mass spectrometry was used for the analysis of methanolic extracts of aerial parts obtained from four Globularia species (G. alypum, G. punctata, G. cordifolia and G. meridionalis). In total, 85 compounds were identified or tentatively identified based on comparison of their retention time, UV and MS