MilliporeSigma
  • Tumor-Localized Secretion of Soluble PD1 Enhances Oncolytic Virotherapy.
Tumor-Localized Secretion of Soluble PD1 Enhances Oncolytic Virotherapy.
Cancer research (2017-03-21)
Mee Y Bartee, Katherine M Dunlap, Eric Bartee
ABSTRACT
Oncolytic virotherapy represents an attractive option for the treatment of a variety of aggressive or refractory tumors. While this therapy is effective at rapidly debulking directly injected tumor masses, achieving complete eradication of established disease has proven difficult. One method to overcome this challenge is to use oncolytic viruses to induce secondary antitumor immune responses. Unfortunately, while the initial induction of these immune responses is typically robust, their subsequent efficacy is often inhibited through a variety of immunoregulatory mechanisms, including the PD1/PDL1 T-cell checkpoint pathway. To overcome this inhibition, we generated a novel recombinant myxoma virus (vPD1), which inhibits the PD1/PDL1 pathway specifically within the tumor microenvironment by secreting a soluble form of PD1 from infected cells. This virus both induced and maintained antitumor CD8
Sigma-Aldrich
Human PDCD1 / Programmed Cell Death Protein 1 ELISA Kit